Fmoc-Val-Cit-PAB-PNP ≥95.0% (by HPLC)

Leverandør: TCI

F1114-100MG F1114-500MG
TCIAF1114-100MGEA 2800 NOK
TCIAF1114-100MG TCIAF1114-500MG
Fmoc-Val-Cit-PAB-PNP ≥95.0% (by HPLC)
Fmoc-Val-Cit-PAB-PNP
Formel: C₄₀H₄₂N₆O₁₀
MW: 766,81 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD22200278
CAS-nummer: 863971-53-3

Bestill nå

Spesifikasjon Testresultater

Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 10.40 to 11.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR