4-Fluoro-3-(trifluoromethyl)benzenesulphonyl chloride ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
4-Fluoro-3-(trifluoromethyl)benzene-1-sulphonyl chloride
F1127-5G
F1127-25G
TCIAF1127-5GEA
1000
NOK
TCIAF1127-5G
TCIAF1127-25G
4-Fluoro-3-(trifluoromethyl)benzenesulphonyl chloride ≥98.0% (by GC)
4-Fluoro-3-(trifluoromethyl)benzenesulphonyl chloride
Formel:
C₇H₃ClF₄O₂S MW: 262,61 g/mol |
MDL nummer:
MFCD01091001 CAS-nummer: 1682-10-6 FN: 3265 ADR: 8,II |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...