2-Fluoro-5-(trifluoromethyl)benzylamine ≥98.0% (by GC)

Leverandør: TCI
Danger

F1148-1G F1148-5G
TCIAF1148-1GEA 675 NOK
TCIAF1148-1G TCIAF1148-5G
2-Fluoro-5-(trifluoromethyl)benzylamine ≥98.0% (by GC)
2-Fluoro-5-(trifluoromethyl)benzylamine
Formel: C₈H₇F₄N
MW: 193,14 g/mol
MDL nummer: MFCD00061162
CAS-nummer: 199296-61-2
FN: 2735
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR