Formamidine Thiocyanate ≥98.0% (by total nitrogen basis)

Leverandør: TCI

Synonymer: Formamidinium Thiocyanate

F1153-1G F1153-5G
TCIAF1153-1GEA 1150 NOK
TCIAF1153-1G TCIAF1153-5G
Formamidine Thiocyanate ≥98.0% (by total nitrogen basis)
Formamidine Thiocyanate
Formel: C₂H₅N₃S
MW: 103,14 g/mol
CAS-nummer: 1821033-48-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystaline
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR