4-Fluorohippuric acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Synonymer:
2-(4-Fluorobenzamido)acetic acid, N-(4-Fluorobenzoyl)glycine
F1162-1G
F1162-200MG
TCIAF1162-1GEA
2025
NOK
TCIAF1162-1G
TCIAF1162-200MG
4-Fluorohippuric acid ≥98.0% (by GC, titration analysis)
4-Fluorohippuric acid
Formel:
C₉H₈FNO₃ MW: 197,17 g/mol |
MDL nummer:
MFCD00462263 CAS-nummer: 366-79-0 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 171.0 to 175.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...