4-Fluoro-N-methoxy-N-methylbenzamide ≥97.0% (by GC)
Leverandør: TCI
F1194-1G
F1194-5G
TCIAF1194-1GEA
310
NOK
TCIAF1194-1G
TCIAF1194-5G
4-Fluoro-N-methoxy-N-methylbenzamide ≥97.0% (by GC)
4-Fluoro-N-methoxy-N-methylbenzamide
Formel:
C₉H₁₀FNO₂ MW: 183,18 g/mol Kokepunkt: 120°C |
MDL nummer:
MFCD02179265 CAS-nummer: 116332-54-8 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...