3-Fluoro-2-methylbenzaldehyde ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-Fluoro-o-tolualdehyde, 3-Fluoro-2-tolualdehyde, 3-Fluoro-o-tolylaldehyde

F1197-1G F1197-5G
TCIAF1197-1GEA 270 NOK
TCIAF1197-1G TCIAF1197-5G
3-Fluoro-2-methylbenzaldehyde ≥98.0% (by GC)
3-Fluoro-2-methylbenzaldehyde
Formel: C₈H₇FO
MW: 138,14 g/mol
Kokepunkt: 60°C
Flash punkt: 81°C
MDL nummer: MFCD00075256
CAS-nummer: 147624-13-3

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 5.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR