2'-Fluoro-5'-nitroacetophenone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-(2-Fluoro-5-nitrophenyl)ethanone

F1199-5G F1199-25G
TCIAF1199-5GEA 850 NOK
TCIAF1199-5G TCIAF1199-25G
2'-Fluoro-5'-nitroacetophenone ≥98.0% (by GC)
2'-Fluoro-5'-nitroacetophenone
Formel: C₈H₆FNO₃
MW: 183,1400 g/mol
MDL nummer: MFCD00277867
CAS-nummer: 79110-05-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR