Fidaxomicin ≥98.0% (by HPLC)

Leverandør: TCI

F1216-100MG F1216-25MG
TCIAF1216-100MGEA 4625 NOK
TCIAF1216-100MG TCIAF1216-25MG
Fidaxomicin ≥98.0% (by HPLC)
Fidaxomicin
Formel: C₅₂H₇₄Cl₂O₁₈
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD27976367
CAS-nummer: 873857-62-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR