2-Fluoro-3-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
F1218-1G
F1218-5G
TCIAF1218-1GEA
340
NOK
TCIAF1218-1G
TCIAF1218-5G
2-Fluoro-3-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Fluoro-3-methoxybenzoic acid
Formel:
C₈H₇FO₃ |
MDL nummer:
MFCD02179621 CAS-nummer: 137654-20-7 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 160.0 to 164.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...