4-Fluoroaniline hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-Fluoroanilinium chloride, 4-Fluorophenylamine hydrochloride, 4-Fluorophenylammonium chloride

F1271-25G F1271-5G
TCIAF1271-25GEA 2500 NOK
TCIAF1271-25G TCIAF1271-5G
4-Fluoroaniline hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Fluoroaniline hydrochloride
Formel: C₆H₆FN·HCl
MW: 147,58 g/mol
Kokepunkt: 167°C
Smeltepunkt: 248°C
MDL nummer: MFCD00035500
CAS-nummer: 2146-07-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR