Diguanidine phosphate ≥95.0% (by titrimetric analysis)

Leverandør: TCI

G0165-25G G0165-500G
TCIAG0165-25GEA 220 NOK
TCIAG0165-25G TCIAG0165-500G
Diguanidine phosphate ≥95.0% (by titrimetric analysis)
Diguanidine phosphate
Formel: C₂H₁₃N₆O₄P
MDL nummer: MFCD00039131
CAS-nummer: 5423-23-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR