Monoethyl glutarate ≥98.0% (by GC, titration analysis)

Leverandør: TCI

Synonymer: 5-Ethoxy-5-oxopentanoic acid, Glutaric Acid Monoethyl Ester

G0271-25G G0271-5G
TCIAG0271-25GEA 1200 NOK
TCIAG0271-25G TCIAG0271-5G
Monoethyl glutarate ≥98.0% (by GC, titration analysis)
Monoethyl glutarate
Formel: C₇H₁₂O₄
MW: 160,17 g/mol
Kokepunkt: 170°C
MDL nummer: MFCD00014379
CAS-nummer: 1070-62-8
EINECS: 213-977-5

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR