Gal[2346Ac]β(1-3)GalN3[46Bzd]-β-MP ≥95.0% (by HPLC)

Leverandør: TCI

Synonymer: 4-Methoxyphenyl 3-O-(2,3,4,6-Tetra-O-acetyl-β-D-galactopyranosyl)-2-azido-4,6-O-benzylidene-2-deoxy-β-D-galactopyranoside

G0330-1G G0330-5G
TCIAG0330-1GEA 3625 NOK
TCIAG0330-1G TCIAG0330-5G
Gal[2346Ac]β(1-3)GalN3[46Bzd]-β-MP ≥95.0% (by HPLC)
Gal[2346Ac]beta(1-3)GalN3[46Bzd]-beta-MP
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8f54cf6df7726f9ac8fb60638efa823a5f07ab6&vid=458f9287c113145b05f838843d2b25aaf4ab0f1a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 191.0 to 195.0 deg-C
Specific rotation [a]20/D +9.0 to +12.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR