Gliquidone ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
N-(Cyclohexylcarbamoyl)-4-(2-(7-methoxy-4,4-dimethyl-1,3-dioxo-3,4-dihydroisoquinolin-2(1H)-yl)ethyl)benzenesulphonamide
G0332-1G
G0332-5G
TCIAG0332-1GEA
690
NOK
TCIAG0332-1G
TCIAG0332-5G
Gliquidone ≥98.0% (by HPLC, titration analysis)
Gliquidone
Formel:
C₂₇H₃₃N₃O₆S MW: 527,64 g/mol |
MDL nummer:
MFCD00631870 CAS-nummer: 33342-05-1 EINECS: 251-463-2 Merck Indeks: 14,04443 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 177.0 to 188.3 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...