Geniposide ≥95.0% (by HPLC)
Leverandør: TCI
Synonymer:
(1S,4aS,7aS)-Methyl 7-(hydroxymethyl)-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate, (1S)-1alpha-(beta-D-Glucopyranosyloxy)-7-(hydroxymethyl)-1,4a alpha,5,7a alpha-tetrahydrocyclopenta[c]pyran-4-carboxylic Acid M, Methyl (1S)-1alpha-(beta-D-Glucopyranosyloxy)-7-(hydroxymethyl)-1,4aalpha,5,7aalpha-tetrahydrocyclopenta[c]pyran-4-carboxylate
Formel:
C₁₇H₂₄O₁₀ MW: 388,37 g/mol |
MDL nummer:
MFCD16036219 CAS-nummer: 24512-63-8 |
Spesifikasjon Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Melting point | 160.0 to 164.0 deg-C |
Specific rotation [a]20/D | +13.0 to +16.0 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...