Granisetron hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 1-Methyl-N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide Hydrochloride

G0401-200MG G0401-1G
TCIAG0401-200MGEA 690 NOK
TCIAG0401-200MG TCIAG0401-1G
Granisetron hydrochloride ≥98.0% (by HPLC, titration analysis)
Granisetron hydrochloride
Formel: C₁₈H₂₄N₄O
MW: 348,88 g/mol
Smeltepunkt: 292°C
MDL nummer: MFCD01747034
CAS-nummer: 107007-99-8
Merck Indeks: 14,04536

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR