GW2580 ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: 5-[3-Methoxy-4-[(4-methoxybenzyl)oxy]benzyl]pyrimidine-2,4-diamine, 5-(3-Methoxy-4-((4-methoxybenzyl)oxy)benzyl)pyrimidine-2,4-diamine

G0504-50MG G0504-250MG
TCIAG0504-50MGEA 985 NOK
TCIAG0504-50MG TCIAG0504-250MG
GW2580 ≥98.0% (by HPLC)
GW2580
Formel: C₂₀H₂₂N₄O₃
MW: 366,42 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD12024701
CAS-nummer: 870483-87-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a10358f93938af8e8c6c1dc3f7cd53fd8ad0919&vid=4a10358f93938af8e8c6c1dc3f7cd53fd8ad0919&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 14.50 to 16.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR