2-Hexyldecanoic acid ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Isopalmitic Acid, Pentadecane-7-carboxylic Acid

H0507-25ML H0507-500ML H0507-100ML
TCIAH0507-25MLEA 230 NOK
TCIAH0507-25ML TCIAH0507-500ML TCIAH0507-100ML
2-Hexyldecanoic acid ≥98.0% (by GC)
2-Hexyldecanoic acid
Formel: C₁₆H₃₂O₂
MW: 256,43 g/mol
Flash punkt: 174°C
MDL nummer: MFCD00059651
CAS-nummer: 25354-97-6
EINECS: 246-885-9
Merck Indeks: 14,04709
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6332d76d61ea1cbcf1226813ad6e5f3bdfc11ba&vid=b6332d76d61ea1cbcf1226813ad6e5f3bdfc11ba&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow to Light orange powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR