4-(Hydroxyamino)quinoline-N-oxide ≥97.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
N-(4-Quinolyl)hydroxylamine 1'-Oxide
H0515-1G
H0515-5G
TCIAH0515-1GEA
1400
NOK
TCIAH0515-1G
TCIAH0515-5G
4-(Hydroxyamino)quinoline-N-oxide ≥97.0% (by titrimetric analysis)
4-(Hydroxyamino)quinoline-N-oxide
Formel:
C₉H₈N₂O₂ MW: 176,18 g/mol Smeltepunkt: 214°C |
MDL nummer:
MFCD00059752 CAS-nummer: 4637-56-3 |
Spesifikasjon Testresultater
Appearance | White to Amber powder to lump |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...