4-(Aminomethyl)benzenesulphonamide hydrochloride ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: alpha-Amino-p-toluenesulphonamide hydrochloride, Homosulphamine hydrochloride, Mafenide hydrochloride, Mafenide HCl

H0522-25G H0522-250G
TCIAH0522-25GEA 1675 NOK
TCIAH0522-25G TCIAH0522-250G
4-(Aminomethyl)benzenesulphonamide hydrochloride ≥98.0% (by HPLC, total nitrogen)
4-(Aminomethyl)benzenesulphonamide hydrochloride
Formel: C₇H₁₁ClN₂O₂S
MW: 222,6924 g/mol
MDL nummer: MFCD00013005
CAS-nummer: 138-37-4
EINECS: 205-325-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR