N-Hydroxy-5-norbornene-2,3-dicarboximide ≥99.0% (by titrimetric analysis) for peptide synthesis
Leverandør: TCI
|
Warning
|
Synonymer:
endo-N-Hydroxybicyclo[2.2.1]hept-5-ene-2,3-dicarboximide
H0528-25G
H0528-250G
TCIAH0528-25GEA
445
NOK
TCIAH0528-25G
TCIAH0528-250G
N-Hydroxy-5-norbornene-2,3-dicarboximide ≥99.0% (by titrimetric analysis) for peptide synthesis
N-Hydroxy-5-norbornene-2,3-dicarboximide
Formel:
C₉H₉NO₃ MW: 179,18 g/mol |
MDL nummer:
MFCD00167583 CAS-nummer: 21715-90-2 EINECS: 244-538-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 167.0 to 171.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...