3-Hydroxy-3-methylvaleric acid ≥95.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-Hydroxy-3-methylpentanoic Acid

H0626-25G H0626-5G
TCIAH0626-25GEA 4025 NOK
TCIAH0626-25G TCIAH0626-5G
3-Hydroxy-3-methylvaleric acid ≥95.0% (by GC, titration analysis)
3-Hydroxy-3-methylvaleric acid
Formel: C₆H₁₂O₃
MW: 132,16 g/mol
Kokepunkt: 144°C
MDL nummer: MFCD00059657
CAS-nummer: 150-96-9

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR