2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
2-(2H-Benzo[d][1,2,3]triazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol
H0716-25G
H0716-500G
TCIAH0716-25GEA
260
NOK
TCIAH0716-25G
TCIAH0716-500G
2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole ≥98.0% (by HPLC, titration analysis)
2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole
Formel:
C₂₀H₂₅N₃O MW: 323,44 g/mol |
MDL nummer:
MFCD00013338 CAS-nummer: 3147-75-9 EINECS: 221-573-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 103.0 to 106.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...