2-Piperidineethanol ≥96.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 2-(2-Hydroxyethyl)piperidine

H0736-25G H0736-500G
TCIAH0736-25GEA 460 NOK
TCIAH0736-25G TCIAH0736-500G
2-Piperidineethanol ≥96.0% (by GC)
2-Piperidineethanol
Formel: C₇H₁₅NO
MW: 129,20 g/mol
Kokepunkt: 234°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00005989
CAS-nummer: 1484-84-0
EINECS: 216-059-2
FN: 1759
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 96.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR