4-Hydroxy-2'-nitrobiphenyl ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4-(2-Nitrophenyl)phenol, 2'-Nitrobiphenyl-4-ol

H0753-1G H0753-5G
TCIAH0753-1GEA 2550 NOK
TCIAH0753-1G TCIAH0753-5G
4-Hydroxy-2'-nitrobiphenyl ≥97.0% (by GC)
4-Hydroxy-2'-nitrobiphenyl
Formel: C₁₂H₉NO₃
MW: 215,21 g/mol
MDL nummer: MFCD00143326
CAS-nummer: 51264-59-6

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 113.0 to 116.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR