4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate ≥97.0% (by GC, titration analysis) catalyst for oxidation free radical

Leverandør: TCI

Synonymer: 2,2,6,6-Tetramethyl-4-(benzoyloxy)piperidine-1-oxyl, 4-Benzoyloxy-TEMPO, 4-Benzoyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Hydroxy-TEMPO Benzoate

H0878-1G H0878-5G
TCIAH0878-1GEA 755 NOK
TCIAH0878-1G TCIAH0878-5G
4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate ≥97.0% (by GC, titration analysis) catalyst for oxidation free radical
4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate
Formel: C₁₆H₂₂NO₃
MW: 276,36 g/mol
MDL nummer: MFCD00075563
CAS-nummer: 3225-26-1

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %
Melting point 102.0 to 106.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR