2-(tert-Butoxycarbonylamino)-1-ethanol ≥98.0% (by GC)
Leverandør: TCI
Warning
Synonymer: N-(2-Hydroxyethyl)carbamic Acid tert-Butyl Ester, 2-(tert-Butoxycarbonyl)ethanolamine, N-(tert-Butoxycarbonyl)ethanolamine, 2-Boc-ethanolamine, tert-Butyl N-(2-Hydroxyethyl)carbamate, 2-(Boc-amino)-1-ethanol, Boc-Glycinol
H0899-25ML H0899-5ML
TCIAH0899-25MLEA 1475 NOK
TCIAH0899-25ML TCIAH0899-5ML
2-(tert-Butoxycarbonylamino)-1-ethanol ≥98.0% (by GC)
2-(tert-Butoxycarbonylamino)-1-ethanol
Formel: C₇H₁₅NO₃
MW: 161,20 g/mol
Kokepunkt: 92°C
MDL nummer: MFCD00056657
CAS-nummer: 26690-80-2
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %