6-Hepten-1-ol ≥96.0% (by GC)

Leverandør: TCI
Warning

H0949-25G H0949-5G
TCIAH0949-25GEA 23000 NOK
TCIAH0949-25G TCIAH0949-5G
6-Hepten-1-ol ≥96.0% (by GC)
6-Hepten-1-ol
Formel: C₇H₁₄O
MW: 114,19 g/mol
Kokepunkt: 113°C
MDL nummer: MFCD00229167
CAS-nummer: 4117-10-6
FN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba000f4127b3a80e6674b2c628726f2475d4f69f&vid=ba000f4127b3a80e6674b2c628726f2475d4f69f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR