(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone ≥94.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
(5S)-(+)-5-(Hydroxymethyl)dihydrofuran-2(3H)-one, (S)-5-(Hydroxymethyl)dihydrofuran-2(3H)-one, (S)-(+)-4,5-Dihydro-5-hydroxymethyl-2(3H)-furanone, (S)-(+)-5-Hydroxymethyltetrahydro-2-furanone
H0958-1G
H0958-5G
TCIAH0958-1GEA
1650
NOK
TCIAH0958-1G
TCIAH0958-5G
(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone ≥94.0% (by GC)
(S)-(+)-gamma-Hydroxymethyl-gamma-butyrolactone
Formel:
C₅H₈O₃ MW: 116,12 g/mol Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD00066224 CAS-nummer: 32780-06-6 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 94.0 % |
Specific rotation [a]20/D | +54.0 to +62.0 deg (C=3, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...