4-Hydroxy-1-cyclohexanecarboxylic acid-δ-lactone ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 3,6-Ethanotetrahydropyran-2-one, 2-Oxabicyclo[2.2.2]octan-3-one

H1143-25G H1143-5G H1143-1G
TCIAH1143-25GEA 8875 NOK
TCIAH1143-25G TCIAH1143-5G TCIAH1143-1G
4-Hydroxy-1-cyclohexanecarboxylic acid-δ-lactone ≥98.0% (by GC)
4-Hydroxy-1-cyclohexanecarboxylic acid-delta-lactone
Formel: C₇H₁₀O₂
MW: 126,16 g/mol
Kokepunkt: 123°C
MDL nummer: MFCD06797111
CAS-nummer: 4350-84-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 130.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR