2-(Hydroxymethyl)phenylboronic acid cyclic monoester ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

H1280-1G H1280-5G
TCIAH1280-1GEA 555 NOK
TCIAH1280-1G TCIAH1280-5G
2-(Hydroxymethyl)phenylboronic acid cyclic monoester ≥97.0% (by HPLC, titration analysis)
2-(Hydroxymethyl)phenylboronic acid cyclic monoester
Formel: C₇H₇BO₂
MDL nummer: MFCD01075677
CAS-nummer: 5735-41-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) 97.0 to 115.0 %
Melting point 98.0 to 102.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR