2-Hydroxy-4-(trifluoromethyl)pyrimidine ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-(Trifluoromethyl)-2-pyrimidinol

H1328-5G H1328-1G
TCIAH1328-5GEA 4000 NOK
TCIAH1328-5G TCIAH1328-1G
2-Hydroxy-4-(trifluoromethyl)pyrimidine ≥97.0% (by GC, titration analysis)
2-Hydroxy-4-(trifluoromethyl)pyrimidine
Formel: C₅H₃F₃N₂O
MW: 164,09 g/mol
MDL nummer: MFCD00192529
CAS-nummer: 104048-92-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a33bd1fa4afcfb59bf110e4704333822c22a259&vid=56f39b7663d61a5812035aab572fa206b5f55340&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 217.0 to 221.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR