Harmol ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 7-Hydroxy-1-methyl-9H-pyrido[3,4-b]indole, 1-Methyl-9H-pyrido[3,4-b]indol-7-ol

H1360-200MG H1360-1G
TCIAH1360-200MGEA 1225 NOK
TCIAH1360-200MG TCIAH1360-1G
Harmol ≥98.0% (by titrimetric analysis)
Harmol
Formel: C₁₂H₁₀N₂O
MW: 198,23 g/mol
Smeltepunkt: 323°C
MDL nummer: MFCD00834164
CAS-nummer: 487-03-6
EINECS: 207-645-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfed60ac7dd12d6427ba5f316bd59009506792d2&vid=cfed60ac7dd12d6427ba5f316bd59009506792d2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR