2'-Hydroxy-5'-nitroacetophenone ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
1-(2-Hydroxy-5-nitrophenyl)ethanone
H1378-5G
H1378-25G
H1378-1G
TCIAH1378-5GEA
2275
NOK
TCIAH1378-5G
TCIAH1378-25G
TCIAH1378-1G
2'-Hydroxy-5'-nitroacetophenone ≥98.0% (by GC, titration analysis)
2'-Hydroxy-5'-nitroacetophenone
Formel:
C₈H₇NO₄ MW: 181,1500 g/mol |
MDL nummer:
MFCD00463816 CAS-nummer: 1450-76-6 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...