3-(Hydroxymethyl)-1-adamantanol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 3-(Hydroxymethyl)adamantan-1-ol, 1-Hydroxy-3-(hydroxymethyl)adamantane

H1415-1G H1415-5G
TCIAH1415-1GEA 445 NOK
TCIAH1415-1G TCIAH1415-5G
3-(Hydroxymethyl)-1-adamantanol ≥98.0% (by GC)
3-(Hydroxymethyl)-1-adamantanol
Formel: C₁₁H₁₈O₂
MW: 182,26 g/mol
MDL nummer: MFCD03105096
CAS-nummer: 38584-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3f580c98963577fc6f7020d9c60bab6f61f0fc2&vid=a3f580c98963577fc6f7020d9c60bab6f61f0fc2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 156.0 to 160.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR