7-(2-Hydroxypropyl)theophylline ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Warning

Synonymer: 7-(2-Hydroxypropyl)-1,3-dimethylxanthine, Proxyphylline, 7-(2-Hydroxypropyl)-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione, 7-(β-Hydroxypropyl)Theophylline

H1430-5G H1430-25G
TCIAH1430-5GEA 485 NOK
TCIAH1430-5G TCIAH1430-25G
7-(2-Hydroxypropyl)theophylline ≥98.0% (by HPLC, total nitrogen)
7-(2-Hydroxypropyl)theophylline
Formel: C₁₀H₁₄N₄O₃
MW: 238,25 g/mol
MDL nummer: MFCD00022833
CAS-nummer: 603-00-9
EINECS: 210-028-7
Merck Indeks: 14,07905
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8549efcb1a1ae2954285bc3800d17509595c1ac1&vid=8549efcb1a1ae2954285bc3800d17509595c1ac1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 134.0 to 138.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR