2-Hydroxybenzaldehyde Phenylhydrazone ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Salicylaldehyde Phenylhydrazone, 2-((2-Phenylhydrazono)methyl)phenol
H1478-5G
H1478-25G
TCIAH1478-5GEA
595
NOK
TCIAH1478-5G
TCIAH1478-25G
2-Hydroxybenzaldehyde Phenylhydrazone ≥98.0% (by GC)
2-Hydroxybenzaldehyde Phenylhydrazone
Formel:
C₁₃H₁₂N₂O MW: 212,25 g/mol Kokepunkt: 234°C |
MDL nummer:
MFCD00136159 CAS-nummer: 614-65-3 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...