2'-Hydroxy-4'-methylacetophenone ≥95.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
1-(2-Hydroxy-4-methylphenyl)ethan-1-one, 1-(2-Hydroxy-4-methylphenyl)ethanone
H1530-5G
H1530-25G
TCIAH1530-5GEA
835
NOK
TCIAH1530-5G
TCIAH1530-25G
2'-Hydroxy-4'-methylacetophenone ≥95.0% (by GC)
2'-Hydroxy-4'-methylacetophenone
Formel:
C₉H₁₀O₂ MW: 150,18 g/mol Kokepunkt: 245°C |
MDL nummer:
MFCD00100559 CAS-nummer: 6921-64-8 |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...