2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Warning

Synonymer: 2-(4,6-Diphenyl-1,3,5-triazin-2-yl)-5-methoxyphenol

H1531-5G H1531-25G
TCIAH1531-5GEA 1100 NOK
TCIAH1531-5G TCIAH1531-25G
2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine
Formel: C₂₂H₁₇N₃O₂
MW: 355,40 g/mol
MDL nummer: MFCD23102325
CAS-nummer: 106556-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6043f8eaf36a39c2cc1b26efa670f9595ea28730&vid=6043f8eaf36a39c2cc1b26efa670f9595ea28730&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 206.0 to 210.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR