4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 2,6,7-Trioxa-1-phosphabicyclo[2.2.2]octane-4-methanol 1-oxide

H1548-1G H1548-5G
TCIAH1548-1GEA 380 NOK
TCIAH1548-1G TCIAH1548-5G
4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide ≥98.0% (by GC)
4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide
Formel: C₅H₉O₅P
MW: 180,096 g/mol
MDL nummer: MFCD01677612
CAS-nummer: 5301-78-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbe18bb893921e2fcb5292ce0d3219d0998df9cf&vid=fbe18bb893921e2fcb5292ce0d3219d0998df9cf&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR