4-(1-Hydroxyethyl)phenol ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
4-Hydroxy-alpha-methylbenzyl alcohol
H1694-200MG
H1694-1G
TCIAH1694-200MGEA
515
NOK
TCIAH1694-200MG
TCIAH1694-1G
4-(1-Hydroxyethyl)phenol ≥98.0% (by GC)
4-(1-Hydroxyethyl)phenol
Formel:
C₈H₁₀O₂ MW: 138,17 g/mol Kokepunkt: 118°C |
MDL nummer:
MFCD00016858 CAS-nummer: 2380-91-8 EINECS: 219-176-7 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 126.0 to 130.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...