1-Methyl-1H-pyrazol-5-ol ≥98.0% (by GC)

Leverandør: TCI
Warning

H1695-1G H1695-5G
TCIAH1695-1GEA 380 NOK
TCIAH1695-1G TCIAH1695-5G
1-Methyl-1H-pyrazol-5-ol ≥98.0% (by GC)
1-Methyl-1H-pyrazol-5-ol
Formel: C₄H₆N₂O
MDL nummer: MFCD05864418
CAS-nummer: 33641-15-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR