5-Hydroxy-3-methyl-2(5H)-furanone ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-Hydroxy-3-methyl-2,5-dihydrofuran-2-one, 5-Hydroxy-3-methylfuran-2(5H)-one

H1747-1G H1747-200MG
TCIAH1747-1GEA 2900 NOK
TCIAH1747-1G TCIAH1747-200MG
5-Hydroxy-3-methyl-2(5H)-furanone ≥95.0% (by GC)
5-Hydroxy-3-methyl-2(5H)-furanone
Formel: C₅H₆O₃
MW: 114,10 g/mol
Kokepunkt: 92°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD15145822
CAS-nummer: 931-23-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR