3-Indoleacrylic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
IAA, 3-(1-H-Indol-3-yl)-2-propenoic acid, 3-(1H-Indol-3-yl)acrylic acid, 3-(3-Indolyl)acrylic Acid
I0025-1G
I0025-5G
TCIAI0025-1GEA
525
NOK
TCIAI0025-1G
TCIAI0025-5G
3-Indoleacrylic acid ≥98.0% (by HPLC, titration analysis)
3-Indoleacrylic acid
Formel:
C₁₁H₉NO₂ MW: 187,20 g/mol |
MDL nummer:
MFCD00005633 CAS-nummer: 1204-06-4 EINECS: 214-872-7 |
Spesifikasjon Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...