Isobutyl vinyl ether ≥99.0% (by GC) stabilised
Leverandør: TCI
|
Danger
|
Stabilisering:
Stabilised with potassium hydroxide
I0099-25ML
I0099-500ML
TCIAI0099-25MLEA
220
NOK
TCIAI0099-25ML
TCIAI0099-500ML
Isobutyl vinyl ether ≥99.0% (by GC) stabilised
Isobutyl vinyl ether
Formel:
C₆H₁₂O MW: 100,16 g/mol Kokepunkt: 83°C Flash punkt: -15°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00008934 CAS-nummer: 109-53-5 EINECS: 203-678-8 FN: 1304 ADR: 3,II |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...