Diisobutylen mixture of isomers ≥94.0% (by GC) stabilised

Leverandør: TCI
Danger

Synonymer: 2,4,4-Trimetylpenten

Stabilisering: stabilisert med BHT/Ionol

I0147-25ML I0147-500ML
TCIAI0147-25MLEA 220 NOK
TCIAI0147-25ML TCIAI0147-500ML
Diisobutylen mixture of isomers ≥94.0% (by GC) stabilised
Diisobutylen
Formel: C₈H₁₆
MW: 112,22 g/mol
Kokepunkt: 105°C
Smeltepunkt: –106°C
Flash punkt: 2°C
MDL nummer: MFCD00008855
CAS-nummer: 25167-70-8
EINECS: 246-690-9
FN: 2050
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf11704306b612da2bff59a5038c5c1c0b8275f9&vid=3747a312c11312949e0c54a3dd52e9052e14ca16&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0% (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR