Tris(2,3-dibromopropyl)isocyanurate ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 1,3,5-Tris(2,3-dibromopropyl)-1,3,5-triazinane-2,4,6-trione, Isocyanuric acid Tris(2,3-dibromopropyl) Ester

I0355-25G I0355-500G
TCIAI0355-25GEA 245 NOK
TCIAI0355-25G TCIAI0355-500G
Tris(2,3-dibromopropyl)isocyanurate ≥97.0% (by titrimetric analysis)
Tris(2,3-dibromopropyl)isocyanurate
Formel: C₁₂H₁₅Br₆N₃O₃
MW: 728,69 g/mol
Smeltepunkt: 110°C
MDL nummer: MFCD00010290
CAS-nummer: 52434-90-9
EINECS: 257-913-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR