1-(Isocyanomethyl)-1H-benzotriazole ≥97.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 1-(Isocyanidomethyl)-1H-benzotriazole, (1H-Benzotriazol-1-yl)methyl Isocyanide

I0455-1G I0455-5G
TCIAI0455-1GEA 1775 NOK
TCIAI0455-1G TCIAI0455-5G
1-(Isocyanomethyl)-1H-benzotriazole ≥97.0% (by HPLC)
1-(Isocyanomethyl)-1H-benzotriazole
Formel: C₈H₆N₄
MW: 158,16 g/mol
MDL nummer: MFCD00961633
CAS-nummer: 87022-42-2

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR