Bis(2-carboxyethyl)isocyanurate ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Warning
Synonymer: Isocyanuric Acid Bis(2-carboxyethyl) Ester
I0584-25G I0584-500G
TCIAI0584-25GEA 395 NOK
TCIAI0584-25G TCIAI0584-500G
Bis(2-carboxyethyl)isocyanurate ≥98.0% (by titrimetric analysis)
Bis(2-carboxyethyl)isocyanurate
Formel: C₉H₁₁N₃O₇
MW: 273,20 g/mol
Smeltepunkt: 289°C
MDL nummer: MFCD04038422
CAS-nummer: 2904-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d558325768c9bab382c4c2f581b211c568d420ea&vid=d558325768c9bab382c4c2f581b211c568d420ea&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in N,N-DMF almost transparency